Assay Details
| Gene Symbol: PLA2G12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243338.1 | NP_001230267.1 | 2 - 3 | 392 | 65 | |
| XM_005671053.3 | XP_005671110.1 | 460 | 65 | |||
| GenBank mRNA | AK345294.1 | - | 2 - 3 | 453 | 65 | |
| AK346162.1 | - | 2 - 3 | 453 | 65 | ||
| EW113750.2 | - | 2 - 3 | 417 | 65 | ||
More Information
Set Membership: |
|

PLA2G12B
Genomic Map