Assay Details
| Gene Symbol: STAMBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243343.1 | NP_001230272.1 | 9 - 10 | 1579 | 62 | |
| XM_021071881.1 | XP_020927540.1 | 1340 | 62 | |||
| XM_021071882.1 | XP_020927541.1 | 3139 | 62 | |||
| XM_021071883.1 | XP_020927542.1 | 1472 | 62 | |||
| GenBank mRNA | AK352067.1 | - | 9 - 10 | 1561 | 62 | |
More Information
Set Membership: |
|

STAMBPL1
Genomic Map