Assay Details
| Gene Symbol: ANXA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243348.1 | NP_001230277.1 | 4 - 5 | 363 | 55 | |
| XM_013997160.2 | XP_013852614.1 | 2075 | 55 | |||
| XM_021088517.1 | XP_020944176.1 | 2355 | 55 | |||
| XM_021088518.1 | XP_020944177.1 | 2108 | 55 | |||
| XM_021088519.1 | XP_020944178.1 | 2077 | 55 | |||
| XM_021088520.1 | XP_020944179.1 | 1963 | 55 | |||
| XM_021088522.1 | XP_020944181.1 | 2353 | 55 | |||
| XM_021088523.1 | XP_020944182.1 | 2353 | 55 | |||
| XM_021088524.1 | XP_020944183.1 | 2352 | 55 | |||
| XM_021088525.1 | XP_020944184.1 | 2354 | 55 | |||
| XM_021088526.1 | XP_020944185.1 | 2355 | 55 | |||
| XM_021088527.1 | XP_020944186.1 | 2355 | 55 | |||
| GenBank mRNA | EW442221.2 | - | 1 - 2 | 85 | 55 | |
| FS673826.1 | - | 3 - 4 | 605 | 55 | ||
More Information
Set Membership: |
|

ANXA9
Genomic Map