Assay Details
| Gene Symbol: ANKRD46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243356.1 | NP_001230285.1 | 3 - 4 | 531 | 99 | |
| XM_005662942.3 | XP_005662999.1 | 559 | 99 | |||
| XM_013996571.2 | XP_013852025.2 | 692 | 99 | |||
| XM_021088530.1 | XP_020944189.1 | 1466 | 99 | |||
| GenBank mRNA | AK344528.1 | - | 3 - 4 | 486 | 99 | |
| AK390426.1 | - | 3 - 4 | 531 | 99 | ||
| FS645670.1 | - | 3 - 4 | 534 | 99 | ||
More Information
Set Membership: |
|

ANKRD46
Genomic Map