Assay Details
| Gene Symbol: ANKRD46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243356.1 | NP_001230285.1 | 4 - 5 | 687 | 66 | |
| XM_005662942.3 | XP_005662999.1 | 715 | 66 | |||
| XM_013996571.2 | XP_013852025.2 | 848 | 66 | |||
| XM_021088530.1 | XP_020944189.1 | 1622 | 66 | |||
| GenBank mRNA | AK344528.1 | - | 4 - 5 | 642 | 66 | |
| AK390426.1 | - | 4 - 5 | 687 | 66 | ||
| FS645670.1 | - | 4 - 5 | 690 | 66 | ||
More Information
Set Membership: |
|

ANKRD46
Genomic Map