Assay Details
| Gene Symbol: MS4A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243358.1 | NP_001230287.1 | 4 - 5 | 647 | 92 | |
| GenBank mRNA | AK231277.1 | - | 4 - 5 | 627 | 92 | |
| AK240438.1 | - | 4 - 5 | 630 | 92 | ||
| CB479849.1 | - | 3 - 4 | 482 | 92 | ||
More Information
Set Membership: |
|

MS4A7
Genomic Map