Assay Details
| Gene Symbol: CHMP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243360.1 | NP_001230289.1 | 5 - 6 | 485 | 65 | |
| GenBank mRNA | AK350235.1 | - | 5 - 6 | 449 | 65 | |
| BP144127.1 | - | 5 - 6 | 485 | 65 | ||
| FD631484.1 | - | 2 - 3 | 285 | 65 | ||
More Information
Set Membership: |
|

CHMP1A
Genomic Map