Assay Details
| Gene Symbol: NDFIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243361.1 | NP_001230290.1 | 1 - 2 | 242 | 91 | |
| GenBank mRNA | AK349550.1 | - | 1 - 2 | 260 | 91 | |
| AK350015.1 | - | 1 - 2 | 242 | 91 | ||
| AK391803.1 | - | 1 - 2 | 261 | 91 | ||
| AK394080.1 | - | 1 - 2 | 240 | 91 | ||
More Information
Set Membership: |
|

NDFIP1
Genomic Map