Assay Details
| Gene Symbol: PEX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243380.1 | NP_001230309.1 | 1 - 2 | 119 | 96 | |
| GenBank mRNA | DT332238.1 | - | 1 - 2 | 119 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243380.1 transcripts.
Set Membership: |
|

PEX2
Genomic Map