Assay Details
| Gene Symbol: RDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243402.1 | NP_001230331.1 | 6 - 7 | 999 | 80 | |
| XM_013989103.2 | XP_013844557.1 | 787 | 80 | |||
| GenBank mRNA | AK343240.1 | - | 5 - 6 | 878 | 80 | |
| AK343418.1 | - | 6 - 7 | 999 | 80 | ||
| AK343426.1 | - | 5 - 6 | 786 | 80 | ||
| AK348008.1 | - | 6 - 7 | 999 | 80 | ||
| AK398111.1 | - | 5 - 6 | 878 | 80 | ||
| AK400729.1 | - | 6 - 7 | 1020 | 80 | ||
| AK400987.1 | - | 6 - 7 | 1000 | 80 | ||
More Information
Set Membership: |
|

RDH12
Genomic Map