Assay Details
| Gene Symbol: MRPL44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243405.1 | NP_001230334.1 | 2 - 3 | 663 | 106 | |
| GenBank mRNA | AK234982.1 | - | 2 - 3 | 640 | 106 | |
| AK235001.1 | - | 2 - 3 | 683 | 106 | ||
| BP164995.1 | - | 2 - 3 | 663 | 106 | ||
| CK467514.1 | - | 1 - 2 | 42 | 106 | ||
More Information
Set Membership: |
|

MRPL44
Genomic Map