Assay Details
| Gene Symbol: HDDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243409.1 | NP_001230338.1 | 3 - 4 | 426 | 95 | |
| GenBank mRNA | AK343282.1 | - | 3 - 4 | 566 | 95 | |
| AK392506.1 | - | 2 - 3 | 728 | 95 | ||
| AK394867.1 | - | 3 - 4 | 528 | 95 | ||
| AK395225.1 | - | 2 - 3 | 703 | 95 | ||
| AK395620.1 | - | 3 - 4 | 426 | 95 | ||
| FS689498.1 | - | 3 - 4 | 426 | 95 | ||
More Information
Set Membership: |
|

HDDC3
Genomic Map