Assay Details
| Gene Symbol: CSDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243420.1 | NP_001230349.1 | 2 - 3 | 424 | 61 | |
| XM_021090925.1 | XP_020946584.1 | 799 | 61 | |||
| XM_021090926.1 | XP_020946585.1 | 528 | 61 | |||
| GenBank mRNA | AK348001.1 | - | 3 - 4 | 535 | 61 | |
More Information
Set Membership: |
|

CSDC2
Genomic Map