Assay Details
| Gene Symbol: TMIGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243436.2 | NP_001230365.1 | 3 - 4 | 275 | 78 | |
| GenBank mRNA | CV869126.1 | - | 2 - 3 | 213 | 78 | |
| CV872063.1 | - | 3 - 4 | 275 | 78 | ||
| CX062116.1 | - | 3 - 4 | 283 | 78 | ||
More Information
Set Membership: |
|

TMIGD3
Genomic Map