Assay Details
| Gene Symbol: C5H12orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243445.1 | NP_001230374.1 | 1 - 2 | 357 | 105 | |
| GenBank mRNA | AK391644.1 | - | 1 - 2 | 323 | 105 | |
| AY610284.1 | - | 1 - 2 | 128 | 105 | ||
| CX061703.1 | - | 1 - 2 | 116 | 105 | ||
| FS665085.1 | - | 1 - 2 | 326 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243445.1 transcripts.
Set Membership: |
|

C5H12orf57
Genomic Map