Assay Details
| Gene Symbol: C5H12orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243445.1 | NP_001230374.1 | 2 - 3 | 527 | 111 | |
| GenBank mRNA | AK391644.1 | - | 2 - 3 | 493 | 111 | |
| AY610284.1 | - | 2 - 3 | 298 | 111 | ||
| CX061703.1 | - | 2 - 3 | 286 | 111 | ||
| FS665085.1 | - | 2 - 3 | 496 | 111 | ||
More Information
Set Membership: |
|

C5H12orf57
Genomic Map