Assay Details
| Gene Symbol: RPL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243458.1 | NP_001230387.1 | 1 - 2 | 41 | 58 | |
| GenBank mRNA | AK348735.1 | - | 1 - 2 | 43 | 58 | |
| AK392102.1 | - | 1 - 2 | 39 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243458.1 transcripts.
Set Membership: |
|

RPL26
Genomic Map