Assay Details
| Gene Symbol: RPL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243458.1 | NP_001230387.1 | 2 - 3 | 208 | 61 | |
| XM_021066165.1 | XP_020921824.1 | 299 | 61 | |||
| GenBank mRNA | AK348735.1 | - | 2 - 3 | 210 | 61 | |
| AK392102.1 | - | 2 - 3 | 206 | 61 | ||
More Information
Set Membership: |
|

RPL26
Genomic Map