Assay Details
| Gene Symbol: C7H6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243459.1 | NP_001230388.1 | 1 - 2 | 171 | 63 | |
| GenBank mRNA | BP165929.1 | - | 1 - 2 | 171 | 63 | |
| DY437202.1 | - | 1 - 2 | 71 | 63 | ||
More Information
Set Membership: |
|

C7H6orf136
Genomic Map