Assay Details
| Gene Symbol: C7H6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243459.1 | NP_001230388.1 | 5 - 6 | 933 | 96 | |
| XM_013977602.2 | XP_013833056.2 | 1050 | 96 | |||
| XM_021098026.1 | XP_020953685.1 | 877 | 96 | |||
| GenBank mRNA | AY610091.1 | - | 4 - 5 | 466 | 96 | |
| DN107093.1 | - | 4 - 5 | 521 | 96 | ||
More Information
Set Membership: |
|

C7H6orf136
Genomic Map