Assay Details
| Gene Symbol: CD5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243468.1 | NP_001230397.1 | 3 - 4 | 523 | 111 | |
| XM_005663237.2 | XP_005663294.1 | 518 | 111 | |||
| GenBank mRNA | AK345954.1 | - | 3 - 4 | 523 | 111 | |
More Information
Set Membership: |
|

CD5L
Genomic Map