Assay Details
| Gene Symbol: RTCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243470.1 | NP_001230399.1 | 4 - 5 | 473 | 63 | |
| GenBank mRNA | AK235362.1 | - | 3 - 4 | 427 | 63 | |
| AK346494.1 | - | 4 - 5 | 473 | 63 | ||
| AK394233.1 | - | 3 - 4 | 473 | 63 | ||
| AK394808.1 | - | 3 - 4 | 496 | 63 | ||
| AY610211.1 | - | 340 | 63 | |||
More Information
Set Membership: |
|

RTCA
Genomic Map