Assay Details
| Gene Symbol: RTRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243494.1 | NP_001230423.1 | 1 - 2 | 156 | 93 | |
| GenBank mRNA | AK240252.1 | - | 1 - 2 | 331 | 93 | |
| AK398046.1 | - | 1 - 2 | 342 | 93 | ||
| AK398721.1 | - | 1 - 2 | 125 | 93 | ||
More Information
Set Membership: |
|

RTRAF
Genomic Map