Assay Details
Gene Symbol: RTRAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243494.1 | NP_001230423.1 | 7 - 8 | 679 | 81 | |
GenBank mRNA | AK240252.1 | - | 7 - 8 | 854 | 81 | |
AK398046.1 | - | 7 - 8 | 865 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
