Assay Details
| Gene Symbol: CHCHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243499.1 | NP_001230428.1 | 2 - 3 | 289 | 71 | |
| GenBank mRNA | AK239789.1 | - | 2 - 3 | 289 | 71 | |
| AY609475.1 | - | 2 - 3 | 212 | 71 | ||
More Information
Set Membership: |
|

CHCHD4
Genomic Map