Assay Details
| Gene Symbol: PIGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243520.1 | NP_001230449.1 | 4 - 5 | 552 | 72 | |
| NM_001243521.1 | NP_001230450.1 | 4 - 5 | 337 | 72 | ||
| NM_001243522.1 | NP_001230451.1 | 4 - 5 | 399 | 72 | ||
| XM_021068231.1 | XP_020923890.1 | 475 | 72 | |||
| GenBank mRNA | AK344057.1 | - | 4 - 5 | 346 | 72 | |
| AK392640.1 | - | 4 - 5 | 392 | 72 | ||
| AK399784.1 | - | 4 - 5 | 548 | 72 | ||
| BM484478.1 | - | 4 - 5 | 336 | 72 | ||
More Information
Set Membership: |
|

PIGP
Genomic Map