Assay Details
| Gene Symbol: MED21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243530.1 | NP_001230459.1 | 1 - 2 | 61 | 87 | |
| GenBank mRNA | AK344778.1 | - | 1 - 2 | 62 | 87 | |
More Information
Set Membership: |
|

MED21
Genomic Map