Assay Details
| Gene Symbol: MRPL40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243559.1 | NP_001230488.1 | 3 - 4 | 384 | 81 | |
| NM_001243560.1 | NP_001230489.1 | 3 - 4 | 409 | 81 | ||
| GenBank mRNA | AK345084.1 | - | 3 - 4 | 359 | 81 | |
| AK391596.1 | - | 3 - 4 | 410 | 81 | ||
| AY609639.1 | - | 3 - 4 | 323 | 81 | ||
| EW093120.2 | - | 3 - 4 | 384 | 81 | ||
| FS664602.1 | - | 3 - 4 | 422 | 81 | ||
More Information
Set Membership: |
|

MRPL40
Genomic Map