Assay Details
| Gene Symbol: MED28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243569.1 | NP_001230498.1 | 3 - 4 | 354 | 113 | |
| GenBank mRNA | AK234158.1 | - | 3 - 4 | 353 | 113 | |
| AK390084.1 | - | 3 - 4 | 353 | 113 | ||
| CJ029267.1 | - | 3 - 4 | 355 | 113 | ||
More Information
Set Membership: |
|

MED28
Genomic Map