Assay Details
| Gene Symbol: MTRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243580.1 | NP_001230509.1 | 8 - 9 | 1198 | 106 | |
| XM_021065101.1 | XP_020920760.1 | 1460 | 106 | |||
| XM_021065102.1 | XP_020920761.1 | 1158 | 106 | |||
| XM_021065103.1 | XP_020920762.1 | 1393 | 106 | |||
More Information
Set Membership: |
|

MTRF1
Genomic Map