Assay Details
| Gene Symbol: INTS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243595.1 | NP_001230524.1 | 11 - 12 | 1446 | 118 | |
| XM_021090951.1 | XP_020946610.1 | 1798 | 118 | |||
| XM_021090952.1 | XP_020946611.1 | 1409 | 118 | |||
| XM_021090953.1 | XP_020946612.1 | 1389 | 118 | |||
| XM_021090954.1 | XP_020946613.1 | 1395 | 118 | |||
| GenBank mRNA | AK343638.1 | - | 11 - 12 | 1446 | 118 | |
More Information
Set Membership: |
|

INTS13
Genomic Map