Assay Details
| Gene Symbol: INTS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243595.1 | NP_001230524.1 | 9 - 10 | 1187 | 95 | |
| XM_021090954.1 | XP_020946613.1 | 1136 | 95 | |||
| GenBank mRNA | AK343638.1 | - | 9 - 10 | 1187 | 95 | |
More Information
Set Membership: |
|

INTS13
Genomic Map