Assay Details
| Gene Symbol: TSNAX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243607.2 | NP_001230536.1 | 4 - 5 | 474 | 64 | |
| GenBank mRNA | AK344359.1 | - | 4 - 5 | 461 | 64 | |
| EW144088.2 | - | 4 - 5 | 515 | 64 | ||
More Information
Set Membership: |
|

TSNAX
Genomic Map