Assay Details
| Gene Symbol: PRKAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243621.1 | NP_001230550.1 | 2 - 3 | 701 | 67 | |
| XM_005670662.3 | XP_005670719.1 | 716 | 67 | |||
| GenBank mRNA | CN158460.1 | - | 1 - 2 | 120 | 67 | |
| DY407095.1 | - | 2 - 3 | 186 | 67 | ||
More Information
Set Membership: |
|

PRKAB1
Genomic Map