Assay Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243624.1 | NP_001230553.1 | 2 - 3 | 285 | 81 | |
| GenBank mRNA | AK351487.1 | - | 2 - 3 | 286 | 81 | |
More Information
Set Membership: |
|

TRMO
Genomic Map