Assay Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243624.1 | NP_001230553.1 | 4 - 5 | 1104 | 84 | |
| GenBank mRNA | AK351487.1 | - | 4 - 5 | 1105 | 84 | |
More Information
Set Membership: |
|

TRMO
Genomic Map