Assay Details
| Gene Symbol: CPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243691.1 | NP_001230620.1 | 4 - 5 | 951 | 63 | |
| GenBank mRNA | AK349998.1 | - | 4 - 5 | 951 | 63 | |
| AK392015.1 | - | 4 - 5 | 986 | 63 | ||
More Information
Set Membership: |
|

CPQ
Genomic Map