Assay Details
Gene Symbol: DRAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243696.1 | NP_001230625.1 | 6 - 7 | 709 | 93 | |
XM_005663558.3 | XP_005663615.1 | 728 | 93 | |||
XM_005663559.3 | XP_005663616.1 | 641 | 93 | |||
XM_021088628.1 | XP_020944287.1 | 583 | 93 | |||
GenBank mRNA | AJ955195.1 | - | 5 - 6 | 585 | 93 | |
EW658272.2 | - | 2 - 3 | 183 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
