Assay Details
| Gene Symbol: NTF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243713.1 | NP_001230642.1 | 2 - 3 | 252 | 88 | |
| XM_013998616.2 | XP_013854070.1 | 198 | 88 | |||
| XM_013998617.2 | XP_013854071.1 | 676 | 88 | |||
| GenBank mRNA | AK347703.1 | - | 2 - 3 | 252 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243713.1, XM_013998616.2, XM_013998617.2 transcripts.
Set Membership: |
|

NTF4
Genomic Map