Assay Details
| Gene Symbol: STX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243720.1 | NP_001230649.1 | 5 - 6 | 697 | 87 | |
| XM_013979910.2 | XP_013835364.1 | 754 | 87 | |||
| XM_021102005.1 | XP_020957664.1 | 755 | 87 | |||
| GenBank mRNA | AK231579.1 | - | 5 - 6 | 697 | 87 | |
| AK236842.1 | - | 5 - 6 | 743 | 87 | ||
More Information
Set Membership: |
|

STX6
Genomic Map