Assay Details
| Gene Symbol: SEPTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243793.1 | NP_001230722.1 | 4 - 5 | 353 | 66 | |
| XM_021074595.1 | XP_020930254.1 | 352 | 66 | |||
| XM_021074596.1 | XP_020930255.1 | 352 | 66 | |||
| XM_021074597.1 | XP_020930256.1 | 567 | 66 | |||
| XM_021074598.1 | XP_020930257.1 | 1665 | 66 | |||
| GenBank mRNA | FD637425.1 | - | 4 - 5 | 353 | 66 | |
| KU358534.1 | - | 4 - 5 | 269 | 66 | ||
More Information
Set Membership: |
|

SEPTIN2
Genomic Map