Assay Details
| Gene Symbol: SEPTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243793.1 | NP_001230722.1 | 7 - 8 | 717 | 102 | |
| XM_021074595.1 | XP_020930254.1 | 716 | 102 | |||
| XM_021074596.1 | XP_020930255.1 | 716 | 102 | |||
| XM_021074597.1 | XP_020930256.1 | 931 | 102 | |||
| XM_021074598.1 | XP_020930257.1 | 2029 | 102 | |||
| XM_021074600.1 | XP_020930259.1 | 647 | 102 | |||
| GenBank mRNA | AK391334.1 | - | 1 - 2 | 89 | 102 | |
| DB794261.1 | - | 1 - 2 | 89 | 102 | ||
| KU358534.1 | - | 7 - 8 | 633 | 102 | ||
More Information
Set Membership: |
|

SEPTIN2
Genomic Map