Assay Details
| Gene Symbol: LARP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243815.2 | NP_001230744.2 | 9 - 10 | 1462 | 98 | |
| GenBank mRNA | CF794019.1 | - | 2 - 3 | 183 | 98 | |
| EW303641.2 | - | 1 - 2 | 183 | 98 | ||
More Information
Set Membership: |
|

LARP7
Genomic Map