Assay Details
| Gene Symbol: SLC7A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243816.1 | NP_001230745.1 | 2 - 3 | 1089 | 76 | |
| XM_021072053.1 | XP_020927712.1 | 2113 | 76 | |||
| XM_021072054.1 | XP_020927713.1 | 2053 | 76 | |||
| GenBank mRNA | AK349273.1 | - | 2 - 3 | 1089 | 76 | |
| AK396238.1 | - | 1 - 2 | 1075 | 76 | ||
| AK400762.1 | - | 2 - 3 | 1087 | 76 | ||
More Information
Set Membership: |
|

SLC7A4
Genomic Map