Assay Details
| Gene Symbol: SLC7A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243816.1 | NP_001230745.1 | 3 - 4 | 1727 | 64 | |
| GenBank mRNA | AK349273.1 | - | 3 - 4 | 1727 | 64 | |
| AK396238.1 | - | 2 - 3 | 1713 | 64 | ||
| AK400762.1 | - | 3 - 4 | 1725 | 64 | ||
More Information
Set Membership: |
|

SLC7A4
Genomic Map