Assay Details
| Gene Symbol: AKR7A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243822.1 | NP_001230751.1 | 5 - 6 | 827 | 67 | |
| GenBank mRNA | AK345819.1 | - | 5 - 6 | 827 | 67 | |
| AK351211.1 | - | 5 - 6 | 845 | 67 | ||
| AK389301.1 | - | 5 - 6 | 840 | 67 | ||
More Information
Set Membership: |
|

AKR7A2
Genomic Map