Assay Details
| Gene Symbol: HSH2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243826.1 | NP_001230755.1 | 2 - 3 | 216 | 58 | |
| XM_021077864.1 | XP_020933523.1 | 2209 | 58 | |||
| GenBank mRNA | AK240093.1 | - | 2 - 3 | 216 | 58 | |
More Information
Set Membership: |
|

HSH2D
Genomic Map