Assay Details
| Gene Symbol: RDH16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243866.1 | NP_001230795.1 | 3 - 4 | 736 | 83 | |
| GenBank mRNA | AK233111.1 | - | 3 - 4 | 736 | 83 | |
| AK233196.1 | - | 3 - 4 | 645 | 83 | ||
| AK233492.1 | - | 3 - 4 | 754 | 83 | ||
| AK400559.1 | - | 3 - 4 | 722 | 83 | ||
More Information
Set Membership: |
|

RDH16
Genomic Map