Assay Details
| Gene Symbol: DPM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244059.1 | NP_001230988.1 | 3 - 4 | 248 | 96 | |
| GenBank mRNA | AK343429.1 | - | 3 - 4 | 242 | 96 | |
| AK345129.1 | - | 3 - 4 | 221 | 96 | ||
| BE014473.1 | - | 3 - 4 | 249 | 96 | ||
More Information
Set Membership: |
|

DPM2
Genomic Map