Assay Details
| Gene Symbol: CDIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244099.1 | NP_001231028.1 | 5 - 6 | 658 | 57 | |
| XM_005662155.3 | XP_005662212.1 | 778 | 57 | |||
| XM_021085649.1 | XP_020941308.1 | 1004 | 57 | |||
| XM_021085650.1 | XP_020941309.1 | 1006 | 57 | |||
| XM_021085651.1 | XP_020941310.1 | 906 | 57 | |||
| XM_021085652.1 | XP_020941311.1 | 875 | 57 | |||
| XM_021085653.1 | XP_020941312.1 | 876 | 57 | |||
| GenBank mRNA | AK389273.1 | - | 1 - 2 | 117 | 57 | |
More Information
Set Membership: |
|

CDIP1
Genomic Map